Gene Transcripts

Gene Symbol
Szt2
Entrez Gene ID
230676
Gene Name
seizure threshold 2
Gene Aliases
6430407N22, ENSMUSG00000058432, TIGR
Chromosome Location
Chr. 4: 118362740 - 118409276 on Build GRCm38
UniGene ID
Mm.415218
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198170.4 NP_937813.3 57 8022
GenBank mRNA AK032181.1 - 5 542
AK129152.1 - 19 3454
FJ998170.1 - 57 8022
GU433214.1 - 57 7945

View Full Product Details