Gene Transcripts
- Gene Symbol
- Hjurp
- Entrez Gene ID
- 381280
- Gene Name
- Holliday junction recognition protein
- Gene Aliases
- 6430706D22Rik, A730008H23Rik, C330011F01Rik, Gm975, Mfleg1
- Chromosome Location
- Chr. 1: 88258856 - 88277633 on Build GRCm38
- UniGene ID
- Mm.384762
- Species
- Mus musculus
- Gene Symbol
- 6430706D22Rik
- Entrez Gene ID
- 78605
- Gene Name
- RIKEN cDNA 6430706D22 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.384762
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK032612.1 |
- |
2 |
2113 |
|
BC062125.1 |
- |
5 |
2521 |
6826662 |
View Full Product Details