Gene Transcripts

Gene Symbol
Hjurp
Entrez Gene ID
381280
Gene Name
Holliday junction recognition protein
Gene Aliases
6430706D22Rik, A730008H23Rik, C330011F01Rik, Gm975, Mfleg1
Chromosome Location
Chr. 1: 88258856 - 88277633 on Build GRCm38
UniGene ID
Mm.384762
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198652.2 NP_941054.2 8 3698
GenBank mRNA BC057309.1 - 8 3639

View Full Product Details