Gene Transcripts

Gene Symbol
CHAF1A
Entrez Gene ID
10036
Gene Name
chromatin assembly factor 1 subunit A
Gene Aliases
CAF-1, CAF1, CAF1B, CAF1P150, P150
Chromosome Location
Chr. 19: 4402596 - 4448322 on Build GRCh38
UniGene ID
Hs.79018
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005483.2 NP_005474.2 6 1376
XM_011527605.2 XP_011525907.1 6 1443
XM_011527606.2 XP_011525908.1 6 1443
XM_011527607.2 XP_011525909.1 5 1386
GenBank mRNA BC067093.1 - 6 1377
U20979.1 - 6 1262

View Full Product Details