Gene Transcripts

Gene Symbol
Drp2
Entrez Gene ID
13497
Gene Name
dystrophin related protein 2
Gene Aliases
AW495265, DRP-2
Chromosome Location
Chr. X: 134404628 - 134456574 on Build GRCm38
UniGene ID
Mm.121662
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010078.3 NP_034208.2 15, 16 2109
GenBank mRNA AK081426.1 - 15, 16 2053
AK158102.1 - 14, 15 1911
BC060605.1 - 13, 14 2688
BC125345.1 - 15, 16 1969 40129860
BC125347.1 - 15, 16 1969 40129862
U43520.1 - 5, 6 715

View Full Product Details