Gene Transcripts

Gene Symbol
Basp1
Entrez Gene ID
70350
Gene Name
brain abundant, membrane attached signal protein 1
Gene Aliases
2610024P12Rik, CAP-23, CAP23, Ckap3, NAP-22, NAP22
Chromosome Location
Chr. 15: 25363277 - 25413764 on Build GRCm38
UniGene ID
Mm.29586
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027395.2 NP_081671.1 2 892
GenBank mRNA AK046868.1 - 2 868
AK159486.1 - 3 283
BC063750.1 - 2 747
BC134388.1 - 2 179

View Full Product Details