Gene Transcripts

Gene Symbol
CAMK4
Entrez Gene ID
814
Gene Name
calcium/calmodulin dependent protein kinase IV
Gene Aliases
CaMK IV, CaMK-GR, caMK
Chromosome Location
Chr. 5: 111223653 - 111498503 on Build GRCh38
UniGene ID
Hs.438801
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323374.1 NP_001310303.1 2 223
NM_001323375.1 NP_001310304.1 2 255
NM_001323376.1 NP_001310305.1 1 371
NM_001323377.1 NP_001310306.1 1 66
NM_001744.5 NP_001735.1 1 371
GenBank mRNA AK311662.1 - 1 226
AK312840.1 - 1 226
BC016695.1 - 1 215
BC025687.1 - 1 213 5205478
BG721313.1 - 1 229
D30742.1 - 1 201
DA396146.1 - 1 359
L17000.1 - 1 165
L24959.1 - 2 223

View Full Product Details