Gene Transcripts

Gene Symbol
Rgl2
Entrez Gene ID
19732
Gene Name
ral guanine nucleotide dissociation stimulator-like 2
Gene Aliases
KE1.5, Rab2l, Rgt2, Rlf
Chromosome Location
Chr. 17: 33929435 - 33937687 on Build GRCm38
UniGene ID
Mm.43777
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009059.2 NP_033085.2 3 397
GenBank mRNA AK144440.1 - 3 396
BC068121.1 - 3 397 6415928
KJ650210.1 - 2 205
U54639.1 - 3 326

View Full Product Details