Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164147.1 | NP_001157619.1 | 19 | 2535 | |
NM_001164148.1 | NP_001157620.1 | 19 | 2532 | ||
NM_001164149.1 | NP_001157621.1 | 19 | 2529 | ||
NM_001164150.1 | NP_001157622.1 | 19 | 2526 | ||
NM_001164151.1 | NP_001157623.1 | 19 | 2523 | ||
NM_001164152.1 | NP_001157624.1 | 19 | 2523 | ||
NM_001164153.1 | NP_001157625.1 | 19 | 2520 | ||
NM_011548.4 | NP_035678.3 | 19 | 2520 | ||
GenBank mRNA | AK014300.1 | - | 3 | 609 | |
AK017617.1 | - | 19 | 2497 | ||
AK034777.1 | - | 19 | 2502 | ||
AK038738.1 | - | 19 | 2510 | ||
AK146786.1 | - | 19 | 2528 | ||
AK147735.1 | - | 18 | 2183 | ||
AK156265.1 | - | 19 | 2512 | ||
AK161009.1 | - | 19 | 2512 | ||
AK170674.1 | - | 19 | 2517 | ||
BC006860.1 | - | 19 | 2474 | 2631291 | |
BC018260.1 | - | 18 | 2157 | 4187386 |