Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_147221.2 | NP_671754.2 | 5 | 1556 | |
GenBank mRNA | AF220434.1 | - | 3 | 1410 | |
AF486579.1 | - | 5 | 1555 | ||
AK145342.1 | - | 2 | 80 | ||
BC066157.1 | - | 3 | 991 | 30468539 | |
JQ043365.1 | - | 4 | 1335 |