Gene Transcripts

Gene Symbol
Lrrc20
Entrez Gene ID
216011
Gene Name
leucine rich repeat containing 20
Gene Aliases
BC036304
Chromosome Location
Chr. 10: 61475276 - 61582791 on Build GRCm38
UniGene ID
Mm.136116
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153542.1 NP_705770.1 5 1460
GenBank mRNA AK136380.1 - 5 1487
AK139384.1 - 5 1493
AK145920.1 - 5 1477
AK150142.1 - 5 1461
AK150246.1 - 5 1461
BC036304.1 - 5 1460 3968534

View Full Product Details