Gene Transcripts

Gene Symbol
Fam84b
Entrez Gene ID
399603
Gene Name
family with sequence similarity 84, member B
Gene Aliases
BF642811, D330050I23Rik
Chromosome Location
Chr. 15: 60818996 - 60825080 on Build GRCm38
UniGene ID
Mm.411968
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162926.1 NP_001156398.1 2 3553
GenBank mRNA AK052388.1 - 1 1430
AK162412.1 - 1 1639

View Full Product Details