Gene Transcripts

Gene Symbol
Wrb
Entrez Gene ID
71446
Gene Name
tryptophan rich basic protein
Gene Aliases
5530402J05Rik, C030018G21Rik, Chd5
Chromosome Location
Chr. 16: 96145354 - 96189247 on Build GRCm38
UniGene ID
Mm.255063
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207301.2 NP_997184.1 2, 3 321
GenBank mRNA AK021091.1 - 2, 3 320
AK172079.1 - 2, 3 334
BC031769.1 - 2, 3 321 4235618

View Full Product Details