Gene Transcripts

Gene Symbol
ARPC1B
Entrez Gene ID
10095
Gene Name
actin related protein 2/3 complex subunit 1B
Gene Aliases
ARC41, p40-ARC, p41-ARC
Chromosome Location
Chr. 7: 99374675 - 99394801 on Build GRCh38
UniGene ID
Hs.489284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005720.3 NP_005711.1 4, 5 509
XM_006715825.1 XP_006715888.1 4, 5 520
XM_006715826.1 XP_006715889.1 4, 5 504
GenBank mRNA AF006084.1 - 4, 5 469
AK314818.1 - 4, 5 454
AU143525.1 - 4, 5 509
AY007111.1 - 2, 3 339
BC002562.2 - 4, 5 442 3138319
BC002988.1 - 4, 5 478 2821335
BC007555.1 - 4, 5 440 2967684

View Full Product Details