Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320778.1 | NP_001307707.1 | 10 | 1415 | |
NM_005697.4 | NP_005688.2 | 9 | 1286 | ||
XM_017021856.1 | XP_016877345.1 | 10 | 1340 | ||
GenBank mRNA | AF005038.2 | - | 9 | 1191 | |
AK096376.1 | - | 7 | 1432 | ||
AK096794.1 | - | 8 | 3075 | ||
AK291884.1 | - | 9 | 1272 | ||
BC001376.2 | - | 9 | 1236 | 3050527 | |
BC004385.1 | - | 9 | 1236 | 3617954 |