Gene Transcripts

Gene Symbol
Heatr5a
Entrez Gene ID
320487
Gene Name
HEAT repeat containing 5A
Gene Aliases
C230081H03Rik, D930036F22Rik, mKIAA1316
Chromosome Location
Chr. 12: 51875871 - 51971333 on Build GRCm38
UniGene ID
Mm.131941
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177171.4 NP_796145.2 3 485
GenBank mRNA AK047221.1 - 3 461
AK053055.1 - 3 486
BC086656.1 - 2 310 30544145

View Full Product Details