Gene Transcripts

Gene Symbol
Ttc16
Entrez Gene ID
338348
Gene Name
tetratricopeptide repeat domain 16
Gene Aliases
1200002K10Rik
Chromosome Location
Chr. 2: 32757026 - 32775663 on Build GRCm38
UniGene ID
Mm.475130
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290563.1 NP_001277492.1 12 1841
NM_001290564.1 NP_001277493.1 12 1896
NM_001290566.1 NP_001277495.1 11 1594
NM_001290567.1 NP_001277496.1 12 1642
NM_177384.3 NP_796358.2 12 1841
GenBank mRNA AK028040.1 - 12 1896
AK089384.1 - 9 2697
AK133273.1 - 12 1619
AK160435.1 - 12 1592
BC140991.1 - 11 1585 9055603
BC145064.1 - 12 1633 9053612

View Full Product Details