Gene Transcripts

Gene Symbol
Zbed4
Entrez Gene ID
223773
Gene Name
zinc finger, BED type containing 4
Gene Aliases
1700009F06Rik
Chromosome Location
Chr. 15: 88751666 - 88784517 on Build GRCm38
UniGene ID
Mm.396622
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181412.3 NP_852077.1 3 1896
GenBank mRNA AK122335.1 - 3 1866
BC052174.1 - 2 1799 6518433
BC092524.1 - 1 1520

View Full Product Details