Gene Transcripts

Gene Symbol
Mtmr6
Entrez Gene ID
219135
Gene Name
myotubularin related protein 6
Gene Aliases
4022440C11Rik, AI428804, AU041072
Chromosome Location
Chr. 14: 60265183 - 60302371 on Build GRCm38
UniGene ID
Mm.247007
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144843.4 NP_659092.1 4, 5 603
GenBank mRNA AB517623.1 - 4, 5 470
AK076218.1 - 4, 5 580
AK154859.1 - 4, 5 523
BC020019.1 - 4, 5 537 3596732

View Full Product Details