Gene Transcripts

Gene Symbol
RABEPK
Entrez Gene ID
10244
Gene Name
Rab9 effector protein with kelch motifs
Gene Aliases
RAB9P40, bA65N13.1, p40
Chromosome Location
Chr. 9: 125200493 - 125234159 on Build GRCh38
UniGene ID
Hs.19012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174152.1 NP_001167623.1 8 970
NM_001174153.1 NP_001167624.1 7 817
NM_005833.3 NP_005824.2 7 1131
XM_005251640.4 XP_005251697.1 7 1043
XM_005251641.4 XP_005251698.1 6 898
XM_005251642.4 XP_005251699.1 6 889
XM_005251644.4 XP_005251701.1 5 739
XM_011518120.2 XP_011516422.1 7 1046
XM_017014177.1 XP_016869666.1 8 955
XM_017014178.1 XP_016869667.1 7 857
XM_017014179.1 XP_016869668.1 6 930
XM_017014180.1 XP_016869669.1 6 704
XM_017014181.1 XP_016869670.1 5 815
XM_017014182.1 XP_016869671.1 6 640
XM_017014183.1 XP_016869672.1 6 977
GenBank mRNA AK290768.1 - 7 975
BC000503.2 - 7 1108 2821471
BC047023.1 - 7 797 5396980
BC053541.1 - 7 940 5724458
BC065725.1 - 8 973 4102355
CR456837.1 - 6 801
CR541851.1 - 6 801
KU178507.1 - 6 801
KU178508.1 - 6 714
KU178509.1 - 5 652
KU178510.1 - 5 648
KU178511.1 - 4 486
Z97074.1 - 8 950

View Full Product Details