Gene Transcripts

Gene Symbol
Aph1a
Entrez Gene ID
226548
Gene Name
anterior pharynx defective 1a homolog (C. elegans)
Gene Aliases
6530402N02Rik, APH-1a
Chromosome Location
Chr. 3: 95893907 - 95898592 on Build GRCm38
UniGene ID
Mm.306954
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146104.3 NP_666216.1 6 2856
NM_146134.2 NP_666246.1 7 2523
GenBank mRNA AK148400.1 - 6 2809
BC012406.1 - 6 2781 3483214

View Full Product Details