Gene Transcripts

Gene Symbol
Cnst
Entrez Gene ID
226744
Gene Name
consortin, connexin sorting protein
Gene Aliases
9630058J23Rik
Chromosome Location
Chr. 1: 179546349 - 179627473 on Build GRCm38
UniGene ID
Mm.87662
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146105.3 NP_666217.2 2 423
GenBank mRNA AK036332.1 - 2 455
AK040666.1 - 2 446
AK040899.1 - 2 454
AK087361.1 - 2 441
AK146681.1 - 2 513
BC054802.1 - 2 423 6837803

View Full Product Details