Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290565.1 | NP_001277494.1 | 4 | 693 | |
NM_153577.2 | NP_705805.1 | 4 | 693 | ||
GenBank mRNA | AA407649.1 | - | 2 | 164 | |
BC004761.1 | - | 4 | 642 | 3155758 | |
BC023803.1 | - | 4 | 680 | 5352953 | |
BC056649.1 | - | 4 | 632 | 5036575 |