Gene Transcripts

Gene Symbol
NAP1L4
Entrez Gene ID
4676
Gene Name
nucleosome assembly protein 1 like 4
Gene Aliases
NAP1L4b, NAP2, NAP2L, hNAP2
Chromosome Location
Chr. 11: 2944430 - 2992377 on Build GRCh38
UniGene ID
Hs.731784
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005969.3 NP_005960.1 4 239
GenBank mRNA AB500094.1 - 2 98
AK299515.1 - 4 162
AK312599.1 - 4 247
AK316242.1 - 4 156
AK316324.1 - 4 183
AK316548.1 - 5 296
BC022090.2 - 4 149 3029455
U77456.1 - 4 247

View Full Product Details