Gene Transcripts

Gene Symbol
Fgfrl1
Entrez Gene ID
116701
Gene Name
fibroblast growth factor receptor-like 1
Gene Aliases
FGFR5, FGFR5beta, FGFR5gamma
Chromosome Location
Chr. 5: 108694229 - 108706950 on Build GRCm38
UniGene ID
Mm.35691
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054071.2 NP_473412.1 3 648
GenBank mRNA AF321300.1 - 3 375
AJ293947.1 - 2 281
BC058745.1 - 2 250 6439421

View Full Product Details