Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_126165.1 | NP_569053.1 | 9 | 1014 | |
GenBank mRNA | AK047821.1 | - | 9 | 1026 | |
AK159117.1 | - | 9 | 1040 | ||
AK169949.1 | - | 9 | 1002 | ||
BC018368.1 | - | 9 | 1014 | 3963907 |