Gene Transcripts
- Gene Symbol
- Enox2
- Entrez Gene ID
- 209224
- Gene Name
- ecto-NOX disulfide-thiol exchanger 2
- Gene Aliases
- APK1, Cova1, tNOX
- Chromosome Location
- Chr. X: 49009707 - 49288245 on Build GRCm38
- UniGene ID
- Mm.327730
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271447.1 |
NP_001258376.1
|
4, 5 |
477 |
|
NM_001271448.1 |
NP_001258377.1
|
3, 4 |
322 |
|
NM_001271449.1 |
NP_001258378.1
|
4, 5 |
466 |
|
NM_001271450.1 |
NP_001258379.1
|
4, 5 |
466 |
|
NM_001271451.1 |
NP_001258380.1
|
4, 5 |
466 |
|
NM_145951.5 |
NP_666063.1
|
4, 5 |
421 |
|
GenBank mRNA |
AK028343.1 |
- |
4, 5 |
467 |
|
AK034058.1 |
- |
4, 5 |
422 |
|
AK041822.1 |
- |
4, 5 |
467 |
|
AK077508.1 |
- |
4, 5 |
448 |
|
AK133956.1 |
- |
3, 4 |
293 |
|
AK137736.1 |
- |
4, 5 |
448 |
|
AK139897.1 |
- |
4, 5 |
448 |
|
BC025915.1 |
- |
4, 5 |
440 |
5043650 |
View Full Product Details