Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172477.4 | NP_766065.1 | 2 | 1151 | |
GenBank mRNA | AK047138.1 | - | 3 | 1139 | |
AK048041.1 | - | 2 | 1135 | ||
AK081176.1 | - | 2 | 1152 | ||
AK160784.1 | - | 2 | 913 | ||
AK165202.1 | - | 2 | 1007 | ||
AK167298.1 | - | 2 | 1005 | ||
BC056367.1 | - | 2 | 1029 | ||
BC057310.1 | - | 2 | 1029 | ||
BC145599.1 | - | 1 | 833 | 9054165 | |
BC150799.1 | - | 1 | 833 | 9087710 |