Gene Transcripts

Gene Symbol
Ncoa7
Entrez Gene ID
211329
Gene Name
nuclear receptor coactivator 7
Gene Aliases
9030406N13Rik
Chromosome Location
Chr. 10: 30645582 - 30805977 on Build GRCm38
UniGene ID
Mm.297903
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172495.5 NP_766083.3 9, 10 2260
GenBank mRNA AK128935.1 - 8, 9 2083
BC042791.1 - 340
BC076623.1 - 7, 8 1866
BC141150.1 - 8, 9 1961 9055762
BC145459.1 - 7, 8 1846

View Full Product Details