Gene Transcripts

Gene Symbol
Scyl1
Entrez Gene ID
78891
Gene Name
SCY1-like 1 (S. cerevisiae)
Gene Aliases
2810011O19Rik, C85140, Ntkl, mdf, mfd, p105
Chromosome Location
Chr. 19: 5758427 - 5771414 on Build GRCm38
UniGene ID
Mm.276063
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023912.2 NP_076401.1 10, 11 1654
GenBank mRNA AF276514.1 - 10, 11 1654
AK019240.1 - 10, 11 1647
AK134356.1 - 10, 11 1653
AK144134.1 - 10, 11 1640
AK145869.1 - 10, 11 1674
AK161813.1 - 10, 11 1653
BC034519.1 - 10, 11 1640 3672524
HM856325.1 - 10, 11 1654
HM856326.1 - 10, 11 1654

View Full Product Details