Gene Transcripts

Gene Symbol
Mtmr14
Entrez Gene ID
97287
Gene Name
myotubularin related protein 14
Gene Aliases
1110061O04Rik, AW553738, C76151
Chromosome Location
Chr. 6: 113237635 - 113281392 on Build GRCm38
UniGene ID
Mm.197816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026849.2 NP_081125.2 19 2448
GenBank mRNA AK004345.1 - 8 1173
AK034867.1 - 18 2309
AK192230.1 - 1 329
AK199499.1 - 1 355
AK204788.1 - 1 353
BB256623.2 - 1 519
BC012436.1 - 17 1976
BC018294.1 - 19 2398 4021243
DV653288.1 - 2 589

View Full Product Details