Gene Transcripts

Gene Symbol
Nfkbiz
Entrez Gene ID
80859
Gene Name
nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, zeta
Gene Aliases
AA408868, INAP, Mail
Chromosome Location
Chr. 16: 55811377 - 55838641 on Build GRCm38
UniGene ID
Mm.247272
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159394.1 NP_001152866.1 5 707
NM_001159395.1 NP_001152867.1 6 589
NM_030612.3 NP_085115.1 7 937
GenBank mRNA AB020974.1 - 7 1149
AB026551.2 - 5 707
AB047549.2 - 6 602
AB196497.1 - 1 42
AK082908.1 - 5 706
AK136509.1 - 2 176
AK141318.1 - 5 706
AK149946.1 - 6 585
AK152016.1 - 5 708
AK156826.1 - 5 708
AK159093.1 - 5 708
AK159171.1 - 5 708
AK159459.1 - 5 708
AK159645.1 - 5 708
AK159917.1 - 5 708
AK160065.1 - 5 706
BC058188.1 - 5 692 5341736

View Full Product Details