Gene Transcripts

Gene Symbol
Gpn2
Entrez Gene ID
100210
Gene Name
GPN-loop GTPase 2
Gene Aliases
AI838661, Atpbd1b, R74630
Chromosome Location
Chr. 4: 133584373 - 133591735 on Build GRCm38
UniGene ID
Mm.274001
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290742.1 NP_001277671.1 5 994
NM_133884.2 NP_598645.2 5 1063
GenBank mRNA BC018407.1 - 5 1056

View Full Product Details