Gene Transcripts

Gene Symbol
Ttll3
Entrez Gene ID
101100
Gene Name
tubulin tyrosine ligase-like family, member 3
Gene Aliases
4833441J24Rik, AI450050
Chromosome Location
Chr. 6: 113389260 - 113414583 on Build GRCm38
UniGene ID
Mm.388573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142732.1 NP_001136204.1 5, 6 779
NM_133923.6 NP_598684.4 5, 6 896
GenBank mRNA AK029462.1 - 5, 6 780
AK080321.1 - 1, 2 310
AM690746.1 - 5, 6 896
BC006830.1 - 3, 4 324
BC021404.1 - 4, 5 528
CF916454.1 - 1, 2 145

View Full Product Details