Gene Transcripts

Gene Symbol
Grwd1
Entrez Gene ID
101612
Gene Name
glutamate-rich WD repeat containing 1
Gene Aliases
A301, AI504353, Grwd, Wdr28
Chromosome Location
Chr. 7: 45825223 - 45830796 on Build GRCm38
UniGene ID
Mm.274847
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153419.2 NP_700468.2 3, 4 475
GenBank mRNA AB001539.1 - 4, 5 547
AK077611.1 - 3, 4 481
BC008121.1 - 3, 4 452
BC028896.1 - 2, 3 344 3674738
BC083143.1 - 3, 4 463
BQ886252.1 - 3, 4 478

View Full Product Details