Gene Transcripts

Gene Symbol
Tmem79
Entrez Gene ID
71913
Gene Name
transmembrane protein 79
Gene Aliases
2310042N02Rik, 2310074C17Rik
Chromosome Location
Chr. 3: 88328653 - 88334433 on Build GRCm38
UniGene ID
Mm.34256
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024246.5 NP_077208.1 4 2134
GenBank mRNA AK009760.1 - 4 2135

View Full Product Details