Gene Transcripts

Gene Symbol
Krtap5-2
Entrez Gene ID
71623
Gene Name
keratin associated protein 5-2
Gene Aliases
4833428E21Rik
Chromosome Location
Chr. 7: 142174532 - 142176005 on Build GRCm38
UniGene ID
Mm.293700
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027844.4 NP_082120.3 1 1403
GenBank mRNA AK014785.1 - 2 870

View Full Product Details