Gene Transcripts

Gene Symbol
Ankle2
Entrez Gene ID
71782
Gene Name
ankyrin repeat and LEM domain containing 2
Gene Aliases
1110001J12Rik, AI661024, D5Ertd585e
Chromosome Location
Chr. 5: 110231004 - 110256651 on Build GRCm38
UniGene ID
Mm.288442
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253814.1 NP_001240743.1 3 1016
NM_027922.2 NP_082198.1 3 1016
GenBank mRNA AK043925.1 - 3 857
AK129192.1 - 2 342
AK132187.1 - 3 1018
AK142497.1 - 1 65
BC065071.1 - 3 979 6410033

View Full Product Details