Gene Transcripts
- Gene Symbol
- ENOX2
- Entrez Gene ID
- 10495
- Gene Name
- ecto-NOX disulfide-thiol exchanger 2
- Gene Aliases
- APK1, COVA1, tNOX
- Chromosome Location
- Chr. X: 130622330 - 130903317 on Build GRCh38
- UniGene ID
- Hs.171458
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281736.1 |
NP_001268665.1
|
9, 10 |
1485 |
|
NM_006375.3 |
NP_006366.2
|
10, 11 |
1629 |
|
NM_182314.2 |
NP_872114.1
|
11, 12 |
1718 |
|
XM_005262354.3 |
XP_005262411.1
|
10, 11 |
1574 |
|
XM_011531244.2 |
XP_011529546.1
|
11, 12 |
1720 |
|
XM_011531247.2 |
XP_011529549.1
|
12, 13 |
1846 |
|
XM_011531249.2 |
XP_011529551.1
|
9, 10 |
1274 |
|
XM_011531251.2 |
XP_011529553.1
|
9, 10 |
1379 |
|
XM_017029226.1 |
XP_016884715.1
|
13, 14 |
1937 |
|
XM_017029227.1 |
XP_016884716.1
|
11, 12 |
1671 |
|
XM_017029228.1 |
XP_016884717.1
|
10, 11 |
1420 |
|
GenBank mRNA |
AF207881.2 |
- |
8, 9 |
1239 |
|
AK000353.1 |
- |
9, 10 |
1402 |
|
AK289812.1 |
- |
11, 12 |
1610 |
|
AK289837.1 |
- |
10, 11 |
1521 |
|
BC108713.1 |
- |
10, 11 |
1506 |
|
BC139781.1 |
- |
8, 9 |
1302 |
|
BC140874.1 |
- |
9, 10 |
1399 |
9021742 |
S72904.1 |
- |
3, 4 |
395 |
|
View Full Product Details