Gene Transcripts

Gene Symbol
Gorasp1
Entrez Gene ID
74498
Gene Name
golgi reassembly stacking protein 1
Gene Aliases
5430411C10Rik, GOLPH5, GRASP65, P65
Chromosome Location
Chr. 9: 119925673 - 119937558 on Build GRCm38
UniGene ID
Mm.104789
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028976.2 NP_083252.1 9 2567
GenBank mRNA AK017293.1 - 2 1597
BC012251.1 - 9 2567 4166619

View Full Product Details