Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029020.2 | NP_083296.2 | 5 | 1044 | |
GenBank mRNA | AK014319.1 | - | 5 | 775 | |
AK014688.1 | - | 5 | 797 | ||
AK030624.1 | - | 5 | 771 | ||
AK038760.1 | - | 5 | 771 | ||
AK049152.1 | - | 5 | 788 | ||
AK076315.1 | - | 5 | 795 | ||
AK166704.1 | - | 5 | 778 | ||
BC015301.1 | - | 5 | 1044 | 4237894 |