Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145824.4 | NP_665823.2 | 7 | 1000 | |
GenBank mRNA | AK173177.1 | - | 7 | 813 | |
AY337314.1 | - | 7 | 1001 | ||
BC024698.1 | - | 7 | 845 | 3667000 | |
EU281316.1 | - | 7 | 861 |