Gene Transcripts
- Gene Symbol
- Dcaf17
- Entrez Gene ID
- 75763
- Gene Name
- DDB1 and CUL4 associated factor 17
- Gene Aliases
- 2810055O12Rik, 4833418A01Rik, A030004A10Rik, A930009G19Rik, AI448937
- Chromosome Location
- Chr. 2: 71055742 - 71114514 on Build GRCm38
- UniGene ID
- Mm.281668
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001165980.1 |
NP_001159452.1
|
1, 2 |
384 |
|
NM_001165981.1 |
NP_001159453.1
|
1, 2 |
384 |
|
NM_001165982.1 |
NP_001159454.1
|
1, 2 |
384 |
|
NM_198005.2 |
NP_932122.2
|
1, 2 |
384 |
|
GenBank mRNA |
AK014717.1 |
- |
1, 2 |
348 |
|
AK032671.1 |
- |
1, 2 |
346 |
|
AK079462.1 |
- |
1, 2 |
178 |
|
AK081839.1 |
- |
1, 2 |
383 |
|
AK160679.1 |
- |
1, 2 |
351 |
|
BC031546.1 |
- |
1, 2 |
276 |
3669736 |
View Full Product Details