Gene Transcripts

Gene Symbol
Dock7
Entrez Gene ID
67299
Gene Name
dedicator of cytokinesis 7
Gene Aliases
3110056M06Rik, Gm430, m
Chromosome Location
Chr. 4: 98936659 - 99122605 on Build GRCm38
UniGene ID
Mm.260623
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290636.1 NP_001277565.1 29, 30 3768
NM_026082.5 NP_080358.3 28, 29 3678
GenBank mRNA BC086672.1 - 9, 10 1186
BC094420.1 - 16, 17 1964
BM964053.1 - 3, 4 430

View Full Product Details