Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026464.2 | NP_080740.2 | 7 | 1459 | |
GenBank mRNA | AK017695.1 | - | 7 | 1433 | |
AK075958.1 | - | 7 | 1459 | ||
AK146093.1 | - | 7 | 1478 | ||
BC011484.1 | - | 5 | 1125 | ||
BC064750.1 | - | 7 | 1418 |