Gene Transcripts

Gene Symbol
Chac2
Entrez Gene ID
68044
Gene Name
ChaC, cation transport regulator 2
Gene Aliases
Chromosome Location
Chr. 11: 30976707 - 30986365 on Build GRCm38
UniGene ID
Mm.32656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290667.1 NP_001277596.1 2 398
NM_026527.3 NP_080803.1 3 434
GenBank mRNA AK007629.1 - 3 358
AK010914.1 - 3 404
AK021391.1 - 2 368
AK078993.1 - 3 420
BC025100.1 - 3 348 3967723

View Full Product Details