Gene Transcripts

Gene Symbol
Ggct
Entrez Gene ID
110175
Gene Name
gamma-glutamyl cyclotransferase
Gene Aliases
A030007L17Rik, AA673177, Gctg, Ggc
Chromosome Location
Chr. 6: 54985095 - 54992965 on Build GRCm38
UniGene ID
Mm.294708
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026637.3 NP_080913.1 4 1304
GenBank mRNA AK028830.1 - 1306
AK076404.1 - 748
AK079465.1 - 4 1306
AK141474.1 - 1354

View Full Product Details