Gene Transcripts

Gene Symbol
Ints2
Entrez Gene ID
70422
Gene Name
integrator complex subunit 2
Gene Aliases
2810417D08Rik, AA408260, AI987735, mKIAA1287
Chromosome Location
Chr. 11: 86210681 - 86257612 on Build GRCm38
UniGene ID
Mm.171304
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027421.2 NP_081697.2 7 1189
GenBank mRNA AK037055.1 - 7 1162
AK129323.1 - 7 1171
BC050081.1 - 7 965 30023969
BC059056.1 - 6 1041 6821329
BC118007.1 - 7 904 40086155

View Full Product Details