Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001267626.1 | NP_001254555.1 | 9 | 958 | |
NM_058212.2 | NP_478119.1 | 9 | 955 | ||
GenBank mRNA | AF362750.1 | - | 9 | 927 | |
AK012970.1 | - | 9 | 963 | ||
BC048572.1 | - | 9 | 838 | 6705809 | |
FJ151218.1 | - | 9 | 930 |