Gene Transcripts

Gene Symbol
GNLY
Entrez Gene ID
10578
Gene Name
granulysin
Gene Aliases
D2S69E, LAG-2, LAG2, NKG5, TLA519
Chromosome Location
Chr. 2: 85694291 - 85698854 on Build GRCh38
UniGene ID
Hs.105806
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302758.1 NP_001289687.1 5 624
NM_006433.4 NP_006424.2 4 543
NM_012483.3 NP_036615.2 5 787
GenBank mRNA AK296106.1 - 2 388
AK304728.1 - 5 557
BC023576.2 - 4 418 4766109
CR541859.1 - 4 415
X05044.1 - 5 647
X54101.1 - 4 543

View Full Product Details